Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger CCHC-type containing 10 | ZCCHC10 | -24.5kb | Anti-sense | Chr. 5 - 132996985 - 133026621 on NCBI Build GRCh38 | |
Downstream | heat shock protein family A (Hsp70) member 4 | HSPA4 |
APG-2, HEL-S-5a, HS24/P52, HSPH2, RY, hsp70, hsp70RY |
468.0 | Sense | Chr. 5 - 133051970 - 133105017 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |