Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000392899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000392899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000466281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000466281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000466281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000470988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000470988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000470988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000623829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000624504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
ENST00000624504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
NM_012369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009514.hg.1 |
NM_012369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000392899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000392899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000466281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000466281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000466281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000470988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000470988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000470988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000623829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000624504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
ENST00000624504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
NM_012369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009514.hg.1 |
NM_012369.2 |
Refseq |