Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8019573 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007114 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007117 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015219 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019700 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015220 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007113 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8007116 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019572 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015222 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015221 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8019570 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8019699 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015218 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8007112 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8007115 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
234631_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11742512_PM_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11742512_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019707.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019707.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019707.hg.1 |
TC17001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019713.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019713.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019713.hg.1 |
TC17001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019708.hg.1 |
ENST00000332991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019708.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019708.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019708.hg.1 |
TC17001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019709.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019709.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019709.hg.1 |
TC17001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019710.hg.1 |
ENST00000332991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019710.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019710.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019710.hg.1 |
TC17001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019712.hg.1 |
ENST00000332991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019712.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019712.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019712.hg.1 |
TC17001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019711.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019711.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019711.hg.1 |
TC17001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206384.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206384.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206384.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206384.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206384.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206384.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206384.hg.1 |
TC1700012405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
ENST00000391413.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012406.hg.1 |
NM_033059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206379.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206379.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206379.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206379.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206379.hg.1 |
TC1700012405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206380.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206380.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206380.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206380.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206380.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206380.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206380.hg.1 |
TC1700012405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012405.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206381.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206381.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206381.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206381.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206381.hg.1 |
TC1700012405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206383.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206383.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206383.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206383.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206383.hg.1 |
TC1700012405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206382.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206382.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206382.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206382.hg.1 |
ENST00000318329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206382.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206382.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206382.hg.1 |
TC1700012405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206385.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206385.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206385.hg.1 |
ENST00000333822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206385.hg.1 |
NM_031960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206385.hg.1 |
TC1700012405.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
234631_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
234631_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234631_at4 |
- |
- |
Human Genome U133 Set |
234631_PM_at |
- |
- |
Human Genome U133 Set |
234631_at |
- |
- |