Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7945665 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7945664 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7945663 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
227886_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
227863_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1564573_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11764253_s_at |
- |
- |
Affymetrix Human Genome U219 Array |
11764253_PM_s_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016010.hg.1 |
TC11003471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016010.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016010.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016010.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016009.hg.1 |
TC11003471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016009.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016009.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016011.hg.1 |
TC11003471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016011.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016011.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016011.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016017.hg.1 |
TC11003471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016017.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016017.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016007.hg.1 |
TC11003471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016007.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016007.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016008.hg.1 |
TC11003471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016008.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016008.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016008.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203259.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203261.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203261.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203261.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203261.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203261.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203261.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203261.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203262.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203269.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203269.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203269.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203269.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203269.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203269.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203269.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203256.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203256.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203256.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203256.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203256.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203256.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203256.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203257.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203257.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203257.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203257.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203257.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203257.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203257.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203255.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203255.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203255.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203255.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203255.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203255.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203255.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203260.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013128.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203254.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203254.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203254.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203254.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203254.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203254.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203254.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000482459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
ENST00000486852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203263.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203270.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203270.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203270.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203270.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203270.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203270.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203270.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203258.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203258.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203258.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203258.hg.1 |
ENST00000340134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203258.hg.1 |
ENST00000382123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203258.hg.1 |
NM_001170820.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203258.hg.1 |
TC1100013128.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
1564573_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1564573_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1564573_at3 |
- |
- |