Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033057.2 | NP_149046.2 | 1 | 556 | 70 | |
GenBank mRNA | BC136881.1 | - | 1 | 556 | 9020872 | 70 |
BC136901.1 | - | 1 | 556 | 9020892 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |