Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005806.hg.1 |
TC09000528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005806.hg.1 |
ENST00000334726.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005806.hg.1 |
NM_001004485.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008289.hg.1 |
ENST00000334726.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008289.hg.1 |
NM_001004485.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008289.hg.1 |
ENST00000334726.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008289.hg.1 |
NM_001004485.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900155525.hg.1 |
TC0900008289.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900155526.hg.1 |
ENST00000334726.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900155526.hg.1 |
NM_001004485.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900155526.hg.1 |
ENST00000334726.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900155526.hg.1 |
NM_001004485.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900155526.hg.1 |
TC0900008289.hg.1 |
Clariom D TC |