Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009509.hg.1 |
ENST00000305045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009509.hg.1 |
NM_001004712.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14009509.hg.1 |
TC14000873.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008576.hg.1 |
ENST00000305045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008576.hg.1 |
NM_001004712.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008576.hg.1 |
ENST00000305045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008576.hg.1 |
NM_001004712.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161506.hg.1 |
ENST00000305045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161506.hg.1 |
NM_001004712.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161506.hg.1 |
ENST00000305045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400161506.hg.1 |
NM_001004712.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400161506.hg.1 |
TC1400008576.hg.1 |
Clariom D TC |