Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005414.hg.1 |
ENST00000302329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005414.hg.1 |
NM_001004727.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005414.hg.1 |
TC11000414.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
ENST00000623137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007516.hg.1 |
NM_001004727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
ENST00000623137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
NM_001004727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
ENST00000623137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
NM_001004727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153519.hg.1 |
TC1100007516.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
ENST00000623137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007516.hg.1 |
NM_001004727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153518.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153518.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153518.hg.1 |
ENST00000302329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153518.hg.1 |
ENST00000624868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153518.hg.1 |
TC1100007516.hg.1 |
Clariom D TC |