Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013593.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013593.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013593.hg.1 |
NM_001005276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025263.hg.1 |
TC07003368.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025263.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025263.hg.1 |
NM_001005276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025262.hg.1 |
TC07003368.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025262.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025262.hg.1 |
NM_001005276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201306.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201306.hg.1 |
NM_001005276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201306.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201306.hg.1 |
NM_001005276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201306.hg.1 |
TC0700013593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013593.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013593.hg.1 |
ENST00000316368.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013593.hg.1 |
NM_001005276.1 |
Refseq |