Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010808.hg.1 |
ENST00000533579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007568.hg.1 |
ENST00000532898.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007570.hg.1 |
ENST00000527276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007521.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007521.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007521.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007521.hg.1 |
NM_001005512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007523.hg.1 |
ENST00000527255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005426.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005426.hg.1 |
NM_001005512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005426.hg.1 |
TC11000418.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100007570.hg.1 |
ENST00000527276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010808.hg.1 |
ENST00000533579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007521.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007521.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007521.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007521.hg.1 |
NM_001005512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007523.hg.1 |
ENST00000527255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153532.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153532.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153532.hg.1 |
TC1100007521.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100153533.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153533.hg.1 |
NM_001005512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153533.hg.1 |
ENST00000446524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153533.hg.1 |
NM_001005512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153533.hg.1 |
TC1100007521.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007568.hg.1 |
ENST00000532898.1 |
Ensembl |