Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000559273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000559273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185728.hg.1 |
TC1500010910.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000559273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000559273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000559776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010910.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000330244.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000558397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000560229.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000560612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000560639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000561068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000561157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
ENST00000561440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185731.hg.1 |
TC1500010910.hg.1 |
Clariom D TC |