Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000526150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000526150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194328.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194328.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194328.hg.1 |
TC1100012285.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000526150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000526150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194325.hg.1 |
TC1100012285.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100194327.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194327.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194327.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194327.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194327.hg.1 |
TC1100012285.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000526150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000526150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012285.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000355430.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000532918.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000540738.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
ENST00000620864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302644.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_001302648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
NM_207429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194324.hg.1 |
TC1100012285.hg.1 |
Clariom D TC |