Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042363.hg.1 |
TC02002643.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042363.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042363.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212211.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212211.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212211.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212211.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212211.hg.1 |
TC0200015330.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
ENST00000342506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015330.hg.1 |
NM_213608.2 |
Refseq |