Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000073.hg.1 |
ENST00000251287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000073.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000073.hg.1 |
TC19000010.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000074.hg.1 |
ENST00000251287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000074.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000074.hg.1 |
TC19000010.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144320.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144320.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144320.hg.1 |
TC1900006471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144320.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144320.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006471.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144316.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144316.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144316.hg.1 |
TC1900006471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144316.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144316.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144318.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144318.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144318.hg.1 |
TC1900006471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144318.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144318.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144319.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144319.hg.1 |
NM_001194.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144319.hg.1 |
TC1900006471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144319.hg.1 |
ENST00000251287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144319.hg.1 |
NM_001194.3 |
Refseq |