Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |