Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005708.hg.1 |
ENST00000299766.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005708.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005708.hg.1 |
TC18000550.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005707.hg.1 |
ENST00000299766.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005707.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005707.hg.1 |
TC18000550.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005709.hg.1 |
ENST00000299766.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005709.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005709.hg.1 |
TC18000550.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008842.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008842.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008842.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160837.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160837.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160837.hg.1 |
TC1800008842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800160837.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160837.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160838.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160838.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160838.hg.1 |
TC1800008842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800160838.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160838.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008842.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008842.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008842.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008842.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160839.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160839.hg.1 |
NM_005912.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160839.hg.1 |
TC1800008842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800160839.hg.1 |
ENST00000299766.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160839.hg.1 |
NM_005912.2 |
Refseq |