Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |