Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010803.hg.1 |
TC05000753.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010803.hg.1 |
ENST00000239446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010803.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013246.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013246.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013246.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194391.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194391.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194391.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194391.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194391.hg.1 |
TC0500013246.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013246.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013246.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013246.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013246.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194392.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194392.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194392.hg.1 |
ENST00000239446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194392.hg.1 |
NM_018930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194392.hg.1 |
TC0500013246.hg.1 |
Clariom D TC |