Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000624994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623741.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623741.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623741.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623741.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623741.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623741.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000623741.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
ENST00000624778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012263.hg.1 |
NR_105056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161678.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161678.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161678.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161678.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161678.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161678.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161678.hg.1 |
TC0500008875.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161677.hg.1 |
TC0500008875.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000194155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000622947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000624994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
ENST00000625033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008875.hg.1 |
NM_018936.3 |
Refseq |