Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013790.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013790.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013790.hg.1 |
TC14001209.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400169394.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169394.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169394.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169394.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169394.hg.1 |
TC1400009395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400169395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169395.hg.1 |
TC1400009395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
ENST00000247225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009395.hg.1 |
NM_030791.3 |
Refseq |