Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018410.hg.1 |
ENST00000378354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018410.hg.1 |
ENST00000539037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018410.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018410.hg.1 |
NM_021006.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018410.hg.1 |
TC17001388.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018413.hg.1 |
ENST00000378354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018413.hg.1 |
ENST00000394484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018413.hg.1 |
ENST00000394486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018413.hg.1 |
ENST00000539037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018413.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018413.hg.1 |
NM_021006.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018413.hg.1 |
TC17001388.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018414.hg.1 |
ENST00000378354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018414.hg.1 |
ENST00000394484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018414.hg.1 |
ENST00000394486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018414.hg.1 |
ENST00000539037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018414.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018414.hg.1 |
NM_021006.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018414.hg.1 |
TC17001388.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700182154.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182154.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182154.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182154.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182154.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182154.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182154.hg.1 |
TC1700010461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182149.hg.1 |
TC1700010461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700182155.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182155.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182155.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182155.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182155.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182155.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182155.hg.1 |
TC1700010461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700182156.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182156.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182156.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182156.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182156.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700182156.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700182156.hg.1 |
TC1700010461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000612839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000616702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
ENST00000619989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010461.hg.1 |
NM_001001437.3 |
Refseq |