Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000738.hg.1 |
ENST00000358134.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000738.hg.1 |
ENST00000416600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000738.hg.1 |
ENST00000428216.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000738.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000738.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000738.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000738.hg.1 |
TC20000043.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
TC2000006561.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145159.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
TC2000006561.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145160.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000416600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
ENST00000428216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_001206491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NM_020746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006561.hg.1 |
NR_037921.1 |
Refseq |