Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001982.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001982.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001982.hg.1 |
TC12000154.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006778.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147620.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147620.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147620.hg.1 |
ENST00000535540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147620.hg.1 |
NM_001190765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147620.hg.1 |
TC1200006778.hg.1 |
Clariom D TC |