Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006765.3 | NP_006756.2 | 6 - 7 | 1146 | 79 | |
NM_178234.2 | NP_839952.1 | 6 - 7 | 1146 | 79 | ||
XM_011544651.2 | XP_011542953.1 | 5 - 6 | 689 | 79 | ||
XM_011544652.2 | XP_011542954.1 | 5 - 6 | 689 | 79 | ||
XM_011544654.2 | XP_011542956.1 | 5 - 6 | 689 | 79 | ||
XM_017013860.1 | XP_016869349.1 | 5 - 6 | 689 | 79 | ||
XM_017013861.1 | XP_016869350.1 | 5 - 6 | 689 | 79 | ||
GenBank mRNA | BC010370.1 | - | 6 - 7 | 1014 | 4334284 | 79 |
BT020002.1 | - | 6 - 7 | 802 | 79 | ||
U42349.1 | - | 6 - 7 | 959 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |