Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057749.2 | NP_477097.1 | 11 - 12 | 1205 | 108 | |
XM_017013957.1 | XP_016869446.1 | 10 - 11 | 1530 | 108 | ||
XM_017013958.1 | XP_016869447.1 | 11 - 12 | 1347 | 108 | ||
XM_017013959.1 | XP_016869448.1 | 11 - 12 | 1449 | 108 | ||
GenBank mRNA | AF091433.1 | - | 10 - 11 | 1138 | 108 | |
AF102778.1 | - | 9 - 10 | 1101 | 108 | ||
AF106690.1 | - | 9 - 10 | 1101 | 108 | ||
AF112857.1 | - | 9 - 10 | 966 | 108 | ||
BC007015.1 | - | 9 - 10 | 940 | 3997513 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |