Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057749.2 | NP_477097.1 | 5 - 6 | 417 | 115 | |
XM_011517366.2 | XP_011515668.1 | 4 - 5 | 742 | 115 | ||
XM_017013957.1 | XP_016869446.1 | 4 - 5 | 742 | 115 | ||
XM_017013958.1 | XP_016869447.1 | 5 - 6 | 559 | 115 | ||
XM_017013959.1 | XP_016869448.1 | 5 - 6 | 661 | 115 | ||
GenBank mRNA | AF091433.1 | - | 4 - 5 | 350 | 115 | |
AF102778.1 | - | 3 - 4 | 313 | 115 | ||
AF106690.1 | - | 3 - 4 | 313 | 115 | ||
AF112857.1 | - | 3 - 4 | 313 | 115 | ||
BC020729.1 | - | 5 - 6 | 364 | 4774554 | 115 | |
BG720611.1 | - | 5 - 6 | 422 | 115 | ||
BJ990268.1 | - | 5 - 6 | 417 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |