Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000631994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000632506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000633742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000634023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
ENST00000634023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
NR_033266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
NR_033266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
NR_033266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009065.hg.1 |
NR_033266.1 |
Refseq |