Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005430.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005430.hg.1 |
TC13000479.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000430708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156149.hg.1 |
ENST00000430708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156149.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156149.hg.1 |
ENST00000430708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156149.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156149.hg.1 |
TC1300008316.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156150.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156150.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156150.hg.1 |
TC1300008316.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156148.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156148.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156148.hg.1 |
TC1300008316.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000430708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
ENST00000577004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008316.hg.1 |
NR_033774.1 |
Refseq |
Human Genome U95 Set |
75785_PM_at |
- |
- |
Human Genome U95 Set |
75785_at |
- |
- |