Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013142.hg.1 |
TC02000884.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013142.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013142.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009449.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009449.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009449.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009449.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013141.hg.1 |
TC02000884.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013141.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013141.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166148.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166148.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166148.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166148.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166148.hg.1 |
TC0200009449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200166149.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166149.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166149.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166149.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166149.hg.1 |
TC0200009449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009449.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009449.hg.1 |
ENST00000437007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009449.hg.1 |
NR_045486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009449.hg.1 |
NR_045486.1 |
Refseq |