Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164367.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164367.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164367.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164367.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164367.hg.1 |
TC1300009635.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000414553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000451662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
ENST00000455958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009635.hg.1 |
NR_047686.1 |
Refseq |