Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270837.1 | NP_001257766.1 | 2 - 3 | 670 | 89 | |
NM_001312.3 | NP_001303.1 | 2 - 3 | 226 | 89 | ||
NR_073082.1 | 2 - 3 | 202 | 89 | |||
NR_073084.1 | 2 - 3 | 979 | 89 | |||
NR_073085.1 | 2 - 3 | 451 | 89 | |||
GenBank mRNA | AK091845.1 | - | 2 - 3 | 979 | 89 | |
AK223251.1 | - | 2 - 3 | 138 | 89 | ||
AK300092.1 | - | 2 - 3 | 586 | 89 | ||
AK311402.1 | - | 2 - 3 | 451 | 89 | ||
AK315757.1 | - | 2 - 3 | 178 | 89 | ||
BC000434.2 | - | 2 - 3 | 157 | 2819982 | 89 | |
BC001931.1 | - | 2 - 3 | 157 | 2819982 | 89 | |
BC034151.1 | - | 2 - 3 | 207 | 4690443 | 89 | |
BC128101.1 | - | 2 - 3 | 181 | 40109449 | 89 | |
BE271979.1 | - | 2 - 3 | 125 | 89 | ||
BT019911.1 | - | 2 - 3 | 138 | 89 | ||
D42123.1 | - | 2 - 3 | 164 | 89 | ||
HY161884.1 | - | 2 - 3 | 202 | 89 | ||
U36190.1 | - | 2 - 3 | 174 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |