Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YLPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK096592.1 | - | 1 | 3059 | 91 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FCF1 rRNA-processing protein | FCF1 |
Bka, C14orf111, CGI-35, UTP24 |
-91.6kb | Anti-sense | Chr. 14 - 74713118 - 74738620 on NCBI Build GRCh38 |
Downstream | prospero homeobox 2 | PROX2 | PROX-2 | 24.0kb | Sense | Chr. 14 - 74853033 - 74876663 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |