Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000303015.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000303015.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000303015.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000303015.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000519923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000519923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000519923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000519923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000520439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000520439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000520439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000520439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000522317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
ENST00000523653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NM_001282534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NM_001282534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NM_001282534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NM_001282534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NM_001282534.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011972.hg.1 |
NR_104210.1 |
Refseq |