Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165090.hg.1 |
TC0400009513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165096.hg.1 |
TC0400009513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009513.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165091.hg.1 |
TC0400009513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000338875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000505610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000513599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
ENST00000515777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
NM_001300781.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
NM_001300782.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165093.hg.1 |
TC0400009513.hg.1 |
Clariom D TC |