Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300818.2 | NP_001287747.1 | 3 - 4 | 332 | 97 | |
NM_001300819.2 | NP_001287748.1 | 2 - 3 | 266 | 97 | ||
XM_011543502.1 | XP_011541804.1 | 3 - 4 | 361 | 97 | ||
GenBank mRNA | BU165290.1 | - | 3 - 4 | 263 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons |