Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004342.3 | NP_001004342.3 | 8 - 9 | 2169 | 91 | |
NM_001300889.1 | NP_001287818.1 | 10 - 11 | 1983 | 91 | ||
XM_011544192.2 | XP_011542494.1 | 8 - 9 | 2163 | 91 | ||
XM_017001323.1 | XP_016856812.1 | 8 - 9 | 2169 | 91 | ||
GenBank mRNA | AK126782.1 | - | 8 - 9 | 2169 | 91 | |
AY253917.1 | - | 10 - 11 | 1941 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons |