Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Hs04398217_m1
See other CNOT8 GE Assays ›
Gene Symbol:
CNOT8
Gene Name:
CCR4-NOT transcription complex subunit 8
Gene Aliases:
CAF1, CALIF, Caf1b, POP2, hCAF1
Chromosome Location:
Chr.5: 154857952 - 154876792 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04398217_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CNOT8
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001301073.1
NP_001288002.1
5 - 6
939
102
NM_001301074.1
NP_001288003.1
4 - 5
777
102
NM_001301075.1
NP_001288004.1
4 - 5
793
102
NM_001301077.1
NP_001288006.1
5 - 6
851
102
NM_001301080.1
NP_001288009.1
4 - 5
750
102
NM_001301082.1
NP_001288011.1
4 - 5
463
102
NM_001301083.1
NP_001288012.1
3 - 4
583
102
NM_001301086.1
NP_001288015.1
3 - 4
588
102
NM_004779.5
NP_004770.4
5 - 6
987
102
XM_005268527.2
XP_005268584.1
5 - 6
1279
102
XM_011537706.1
XP_011536008.1
6 - 7
961
102
XM_011537708.2
XP_011536010.1
4 - 5
464
102
XM_017010048.1
XP_016865537.1
6 - 7
846
102
XM_017010049.1
XP_016865538.1
6 - 7
1441
102
XM_017010050.1
XP_016865539.1
6 - 7
1426
102
XM_017010051.1
XP_016865540.1
6 - 7
1476
102
XM_017010052.1
XP_016865541.1
5 - 6
1231
102
XM_017010053.1
XP_016865542.1
5 - 6
1249
102
XM_017010054.1
XP_016865543.1
4 - 5
770
102
XM_017010055.1
XP_016865544.1
4 - 5
802
102
XM_017010056.1
XP_016865545.1
4 - 5
888
102
XM_017010057.1
XP_016865546.1
6 - 7
1004
102
XM_017010058.1
XP_016865547.1
3 - 4
592
102
GenBank mRNA
AF087844.1
-
4 - 5
689
102
AF180476.1
-
4 - 5
689
102
AK025358.1
-
5 - 6
908
102
AK297381.1
-
4 - 5
658
102
AK299589.1
-
6 - 7
819
102
AK303775.1
-
4 - 5
644
102
AK309359.1
-
5 - 6
741
102
AK314317.1
-
5 - 6
939
102
AK316055.1
-
3 - 4
568
102
AK316221.1
-
4 - 5
463
102
AK316269.1
-
3 - 4
581
102
AL122045.1
-
5 - 6
868
102
BC017366.2
-
5 - 6
798
3506054
102
BT006857.1
-
4 - 5
617
102
BX647124.1
-
3 - 4
330
102
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Recommended Assay
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene CNOT8
Panther Classification:
Molecular Function -
mRNA polyadenylation factor
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA poly(A) tail shortening
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
positive regulation of cell proliferation
negative regulation of cell proliferation
gene silencing by miRNA
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
positive regulation of mRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
transcription factor activity, sequence-specific DNA binding
RNA binding
poly(A)-specific ribonuclease activity
protein binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0