Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
ENST00000396613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_001287010.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_001287011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_001287012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_022343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
ENST00000396613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_001287010.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_001287011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_001287012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NM_022343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
NR_104641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148445.hg.1 |
TC0900007115.hg.1 |
Clariom D TC |