Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304548.1 | NP_001291477.1 | 19 - 20 | 3237 | 71 | |
XM_017029452.1 | XP_016884941.1 | 19 - 20 | 3230 | 71 | ||
XM_017029453.1 | XP_016884942.1 | 19 - 20 | 3230 | 71 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAGE family member B16 | MAGEB16 | -193.6kb | Sense | Chr. X - 35798317 - 35803753 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928627 | LOC101928627 | 365.2kb | Anti-sense | Chr. X - 36365626 - 36440292 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |