Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110523.1 | 4 - 5 | 724 | 66 | ||
GenBank mRNA | BC033837.1 | - | 4 - 5 | 697 | 66 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105373085 | LOC105373085 | -362.1kb | Sense | Chr. 22 - 49047480 - 49052405 on NCBI Build GRCh38 | |
Downstream | microRNA 3667 | MIR3667 | 126.7kb | Sense | Chr. 22 - 49543393 - 49543466 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |