Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177108.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177108.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177108.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177108.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177108.hg.1 |
TC0700010843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700177106.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177106.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177106.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177106.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177106.hg.1 |
TC0700010843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700177105.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177105.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177105.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177105.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177105.hg.1 |
TC0700010843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700177107.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177107.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177107.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177107.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177107.hg.1 |
TC0700010843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
ENST00000443406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010843.hg.1 |
NR_110831.1 |
Refseq |