Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111932.1 | 19 - 20 | 2506 | 72 | ||
GenBank mRNA | BC110287.1 | - | 19 - 20 | 2436 | 5165262 | 72 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928553 | LOC101928553 | -93.1kb | Anti-sense | Chr. 2 - 148875995 - 148890572 on NCBI Build GRCh38 | |
Downstream | LY6/PLAUR domain containing 6B | LYPD6B |
CT116, LYPD7 |
11.7kb | Sense | Chr. 2 - 149038467 - 149215262 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |