Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154898.hg.1 |
TC0400007978.hg.1 |
Clariom D TC |