Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXG1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125758.1 | 6 - 7 | 778 | 75 | ||
GenBank mRNA | BX426122.2 | - | 6 - 7 | 75 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC728755 | LOC728755 | -1.1mb | Sense | Chr. 14 - 27315817 - 27673221 on NCBI Build GRCh38 | |
Downstream | forkhead box G1 | FOXG1 |
BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN |
37.8kb | Anti-sense | Chr. 14 - 28767072 - 28770277 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons |