Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012372.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012372.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012372.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012372.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012372.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012372.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184379.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184379.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184379.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184379.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184379.hg.1 |
TC1000012372.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000184378.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184378.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184378.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184378.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184378.hg.1 |
TC1000012372.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000184380.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184380.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184380.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184380.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184380.hg.1 |
TC1000012372.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012372.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012372.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012372.hg.1 |
ENST00000423232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012372.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012372.hg.1 |
NR_125763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012372.hg.1 |
NR_125763.1 |
Refseq |