Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215017.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215017.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215017.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215017.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215017.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215017.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215017.hg.1 |
TC0100015394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100215018.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215018.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215018.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215018.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215018.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215018.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215018.hg.1 |
TC0100015394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000423907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000430316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000434879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000437308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
ENST00000443219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125972.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015394.hg.1 |
NR_125973.1 |
Refseq |