Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000505467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000505467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000505467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000505467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
ENST00000511287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103788.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012276.hg.1 |
NR_103789.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191852.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191852.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191852.hg.1 |
ENST00000506660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191852.hg.1 |
NR_103787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191852.hg.1 |
TC0300012276.hg.1 |
Clariom D TC |