Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000181.3 | NP_000172.2 | 12 | 2202 | 120 | |
NM_001284290.1 | NP_001271219.1 | 10 | 1764 | 120 | ||
NM_001293104.1 | NP_001280033.1 | 11 | 2017 | 120 | ||
NM_001293105.1 | NP_001280034.1 | 10 | 1874 | 120 | ||
NR_120531.1 | 11 | 2117 | 120 | |||
XM_005250297.3 | XP_005250354.1 | 11 | 2044 | 120 | ||
XM_011516113.1 | XP_011514415.1 | 11 | 1946 | 120 | ||
XM_011516114.1 | XP_011514416.1 | 11 | 1874 | 120 | ||
XM_017012091.1 | XP_016867580.1 | 10 | 1793 | 120 | ||
XM_017012092.1 | XP_016867581.1 | 10 | 1780 | 120 | ||
XM_017012093.1 | XP_016867582.1 | 10 | 1721 | 120 | ||
GenBank mRNA | AA947121.1 | - | 1 | 112 | 120 | |
AK096764.1 | - | 4 | 2460 | 120 | ||
AK223406.1 | - | 12 | 2101 | 120 | ||
AK300223.1 | - | 12 | 1991 | 120 | ||
BC014142.2 | - | 12 | 2072 | 4662011 | 120 | |
BM719712.1 | - | 3 | 455 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |