Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131177.1 | 1 - 2 | 350 | 82 | ||
GenBank mRNA | AW796546.1 | - | 1 - 2 | 84 | 82 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | heat shock factor binding protein 1 like 1 | HSBP1L1 | -2.0kb | Anti-sense | Chr. 18 - 79964582 - 79970822 on NCBI Build GRCh38 | |
Downstream | ribosome binding factor A (putative) | RBFA |
C18orf22, HsT169 |
45.8kb | Anti-sense | Chr. 18 - 80034346 - 80050652 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons |