Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036033.hg.1 |
TC02002218.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036033.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036033.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202057.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202057.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202057.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202057.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202057.hg.1 |
TC0200013913.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
ENST00000263339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013913.hg.1 |
NM_000575.4 |
Refseq |